Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021435.3 | NP_067410.1 | 9, 10 | 939 | ||
GenBank mRNA | AB041549.1 | - | 9, 10 | 940 | ||
AK029046.1 | - | 8, 9 | 3560 | |||
AK029969.1 | - | 9, 10 | 997 | |||
AK033913.1 | - | 9, 10 | 869 | |||
AK042150.1 | - | 9, 10 | 839 | |||
AK045791.1 | - | 9, 10 | 882 | |||
AK138591.1 | - | 9, 10 | 880 | |||
AK163299.1 | - | 4, 5 | 407 | |||
BC016275.1 | - | 9, 10 | 870 | |||
BC033512.1 | - | 9, 10 | 864 | 4017336 |