Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163354.1 | NP_001156826.1 | 20 | 2123 | ||
NM_001163355.1 | NP_001156827.1 | 20 | 2150 | |||
NM_021536.7 | NP_067511.4 | 19 | 2027 | |||
GenBank mRNA | AF244542.1 | - | 6 | 661 | ||
AK019059.1 | - | 19 | 1987 | |||
AK028597.1 | - | 19 | 1970 | |||
AK031139.1 | - | 17 | 1689 | |||
AK032096.1 | - | 19 | 2000 | |||
AK041119.1 | - | 20 | 2059 | |||
AK054472.1 | - | 19 | 1998 | |||
AK078955.1 | - | 19 | 1970 | |||
AK090071.1 | - | 12 | 1354 | |||
AK138830.1 | - | 19 | 1968 | |||
AK141621.1 | - | 20 | 1977 | |||
AK147749.1 | - | 20 | 2102 | |||
AK147801.1 | - | 20 | 2128 | |||
AK167502.1 | - | 20 | 2150 | |||
AY974596.1 | - | 2 | 212 | |||
BC006918.1 | - | 11 | 1241 | |||
BC025021.1 | - | 6 | 1263 | |||
BC044301.1 | - | 9 | 989 | |||
BC046785.1 | - | 19 | 2010 | 6416127 | ||
BC052882.1 | - | 10 | 1067 | |||
BC058350.1 | - | 19 | 1942 | 6815832 |