Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nisch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022656.2 | NP_073147.2 | 21 | 4835 | ||
GenBank mRNA | AF144133.1 | - | 2 | 516 | ||
AF315344.1 | - | 17 | 4288 | |||
AK122412.1 | - | 19 | 4333 | |||
AY032852.1 | - | 4 | 970 | |||
BC089329.1 | - | 16 | 4054 | |||
BC108364.1 | - | 16 | 4029 | |||
BC144980.1 | - | 17 | 4164 |