Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164466.1 | NP_001157938.1 | 8 | 1370 | ||
NM_022722.3 | NP_073559.3 | 8 | 1370 | |||
GenBank mRNA | AF227731.1 | - | 8 | 1288 | ||
AF249296.1 | - | 8 | 1357 | |||
AK004822.1 | - | 8 | 1370 | |||
AK004899.2 | - | 8 | 1367 | |||
BC029718.1 | - | 6 | 709 | |||
BC037086.1 | - | 8 | 1339 | 5123860 |