Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164885.1 | NP_001158357.1 | 12 | 1874 | ||
NM_022882.4 | NP_075020.2 | 12 | 1750 | |||
GenBank mRNA | AF286723.1 | - | 12 | 1750 | ||
BC039698.1 | - | 12 | 1863 | 5101211 |