Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023049.1 | NP_075536.1 | 7 | 1344 | ||
GenBank mRNA | AB221595.1 | - | 6 | 954 | ||
AF155353.1 | - | 5 | 811 | |||
AK083400.1 | - | 5 | 870 | |||
AK155235.1 | - | 3 | 1058 | |||
BC031161.1 | - | 7 | 1344 | 4981777 |