Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023053.3 | NP_075540.1 | 5 | 686 | ||
GenBank mRNA | AF292033.1 | - | 625 | |||
AF295097.1 | - | 4 | 544 | |||
AJ297390.1 | - | 5 | 636 | |||
AK018524.1 | - | 1 | 53 | |||
AK033504.1 | - | 5 | 645 | |||
AK049646.1 | - | 5 | 680 | |||
AK203652.1 | - | 2 | 91 | |||
BC004850.1 | - | 5 | 641 | 2810960 |