Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025446.3 | NP_079722.1 | 4 | 497 | ||
GenBank mRNA | AK005332.1 | - | 4 | 525 | ||
AK008210.1 | - | 4 | 500 | |||
AK162306.1 | - | 4 | 516 | |||
BC035305.1 | - | 4 | 647 | 3980923 | ||
EF410113.1 | - | 2 | 382 |