Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025446.3 | NP_079722.1 | 5 | 683 | ||
GenBank mRNA | AK005332.1 | - | 5 | 711 | ||
AK008210.1 | - | 5 | 686 | |||
AK162306.1 | - | 5 | 702 | |||
BC035305.1 | - | 5 | 833 | 3980923 | ||
EF410113.1 | - | 3 | 562 |