Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290807.1 | NP_001277736.1 | 5, 6 | 479 | ||
NM_001290808.1 | NP_001277737.1 | 4, 5 | 509 | |||
NM_025475.3 | NP_079751.1 | 5, 6 | 579 | |||
GenBank mRNA | AK007106.1 | - | 5, 6 | 481 | ||
AK010445.1 | - | 5, 6 | 526 | |||
AK012086.1 | - | 5, 6 | 578 | |||
AK012779.1 | - | 4, 5 | 509 | |||
AK012960.1 | - | 5, 6 | 526 | |||
AK132300.1 | - | 5, 6 | 535 | |||
BC086677.1 | - | 5, 6 | 517 | 30660268 |