Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025570.2 | NP_079846.1 | 1, 2 | 115 | ||
GenBank mRNA | AB049645.1 | - | 1, 2 | 88 | ||
AK004053.1 | - | 1, 2 | 100 | |||
AK011341.1 | - | 1, 2 | 116 | |||
AK015199.1 | - | 99 | ||||
AK168665.1 | - | 1, 2 | 89 | |||
BC039985.1 | - | 1, 2 | 85 | 1244957 |