Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025579.3 | NP_079855.2 | 2 | 215 | ||
GenBank mRNA | AK007811.1 | - | 2 | 178 | ||
AK013133.1 | - | 2 | 178 | |||
AK132927.1 | - | 2 | 216 | |||
AK139748.1 | - | 2 | 129 | |||
BC019668.1 | - | 2 | 160 | 3493047 | ||
BY069348.1 | - | 2 | 216 |