Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025609.2 | NP_079885.2 | 6 | 602 | ||
GenBank mRNA | BC027054.1 | - | 6 | 594 | ||
BC041110.1 | - | 6 | 597 | |||
BC054369.1 | - | 6 | 602 | 5356886 | ||
BC114637.1 | - | 6 | 623 |