Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad2l1bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025649.3 | NP_079925.2 | 2 | 124 | ||
GenBank mRNA | AK002377.1 | - | 2 | 110 | ||
AK004224.1 | - | 2 | 78 | |||
AK156569.1 | - | 2 | 127 | |||
AK220322.1 | - | 2 | 93 | |||
BC005768.1 | - | 2 | 82 | 2649017 |