Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025692.3 | NP_079968.2 | 9 | 1073 | ||
GenBank mRNA | AK017787.1 | - | 9 | 1073 | ||
AK077825.1 | - | 9 | 1054 | |||
AK151823.1 | - | 9 | 1030 | |||
AK152029.1 | - | 9 | 1068 | |||
AK168907.1 | - | 9 | 1067 | |||
AK178397.1 | - | 2 | 164 |