Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025770.3 | NP_080046.3 | 6 | 548 | ||
GenBank mRNA | AB079383.1 | - | 5 | 504 | ||
AB091691.1 | - | 5 | 525 | |||
AK009076.1 | - | 6 | 532 | |||
AK017355.1 | - | 6 | 530 | |||
AK077333.1 | - | 6 | 521 | |||
BC023455.1 | - | 5 | 524 | 5249268 |