Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173547.2 | NP_001167018.1 | 6 | 761 | ||
NM_001242615.2 | NP_001229544.1 | 6 | 761 | |||
NM_001242616.2 | NP_001229545.1 | 6 | 761 | |||
NM_001290738.1 | NP_001277667.1 | 5 | 680 | |||
NM_181516.6 | NP_852657.1 | 6 | 761 | |||
GenBank mRNA | AK004035.1 | - | 6 | 718 | ||
AK075615.1 | - | 6 | 718 | |||
AK076481.1 | - | 6 | 729 | |||
AK160327.1 | - | 6 | 730 | |||
AK170662.1 | - | 6 | 730 | |||
AK178858.1 | - | 3 | 179 | |||
AK183640.1 | - | 2 | 61 | |||
AK188231.1 | - | 4 | 219 | |||
AY231459.1 | - | 6 | 480 | |||
AY231460.1 | - | 6 | 480 | |||
AY258040.1 | - | 6 | 480 | |||
AY258041.1 | - | 6 | 480 | |||
BC015305.1 | - | 6 | 726 | 4221041 | ||
CD807936.1 | - | 5 | 626 | |||
EF524069.1 | - | 5 | 360 |