Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024434.6 | NP_077754.3 | 5 | 551 | ||
GenBank mRNA | AF334160.1 | - | 5 | 512 | ||
AK002819.1 | - | 5 | 487 | |||
AK005334.1 | - | 5 | 464 | |||
AK008600.1 | - | 5 | 438 | |||
AK010384.1 | - | 5 | 436 | |||
AK010502.1 | - | 5 | 504 | |||
AK011604.1 | - | 5 | 437 | |||
AK089023.1 | - | 5 | 518 | |||
AK166958.1 | - | 5 | 435 | |||
AK169032.1 | - | 5 | 437 | |||
AK192064.1 | - | 1 | 22 | |||
AK217131.1 | - | 1 | 22 | |||
BC016536.1 | - | 5 | 551 | 3488469 | ||
CT010237.1 | - | 5 | 465 | |||
CT010301.1 | - | 5 | 465 |