Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163687.1 | NP_001157159.1 | 2, 3 | 431 | ||
NM_025972.4 | NP_080248.2 | 2, 3 | 437 | |||
GenBank mRNA | AB162194.1 | - | 2, 3 | 403 | ||
AK008776.1 | - | 2, 3 | 439 | |||
AK017141.1 | - | 2, 3 | 416 | |||
AK133485.1 | - | 2, 3 | 422 | |||
AK133658.1 | - | 2, 3 | 439 | |||
AK146191.1 | - | 2, 3 | 426 | |||
BC004572.1 | - | 2, 3 | 407 | 3586255 |