Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290500.1 | NP_001277429.1 | 2 | 269 | ||
NM_001290502.1 | NP_001277431.1 | 2 | 507 | |||
NM_025978.4 | NP_080254.2 | 2 | 269 | |||
NM_027619.4 | NP_081895.2 | 2 | 269 | |||
GenBank mRNA | AK012246.1 | - | 2 | 231 | ||
AK015308.1 | - | 2 | 229 | |||
AK016619.1 | - | 2 | 189 | |||
AK031605.1 | - | 2 | 500 | |||
AK035570.1 | - | 2 | 239 | |||
AK041095.1 | - | 2 | 243 | |||
AK044266.1 | - | 2 | 244 | |||
AK163444.1 | - | 2 | 269 | |||
AK173315.1 | - | 2 | 239 | |||
BC021448.1 | - | 2 | 277 | |||
BC024847.1 | - | 2 | 234 | 5355806 | ||
BC138212.1 | - | 2 | 231 | 8861231 | ||
BC145944.1 | - | 2 | 231 | 40131247 |