Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mastl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025979.4 | NP_080255.3 | 3 | 517 | ||
GenBank mRNA | AK030140.1 | - | 3 | 412 | ||
BC103779.1 | - | 3 | 393 | 30480071 | ||
CN536399.1 | - | 3 | 290 |