Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026029.3 | NP_080305.2 | 1 | 110 | ||
GenBank mRNA | AK007439.1 | - | 1 | 80 | ||
AK010992.1 | - | 1 | 68 | |||
AK012569.1 | - | 1 | 88 | |||
AK013212.1 | - | 1 | 88 | |||
AK138073.1 | - | 87 | ||||
AK138290.1 | - | 1 | 75 | |||
BC061012.1 | - | 1 | 102 | 6587019 | ||
BY122171.1 | - | 1 | 110 |