Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038696.1 | NP_001033785.1 | 3 | 366 | ||
NM_001287015.1 | NP_001273944.1 | 3 | 366 | |||
NM_026043.3 | NP_080319.2 | 3 | 366 | |||
GenBank mRNA | AK013288.1 | - | 3 | 310 | ||
AK134241.1 | - | 3 | 331 | |||
AK161063.1 | - | 3 | 349 | |||
AK161974.1 | - | 3 | 319 | |||
AK163729.1 | - | 3 | 319 | |||
AK173272.1 | - | 3 | 117 | |||
BC043695.1 | - | 3 | 335 | 5250774 | ||
BY004586.1 | - | 3 | 367 |