Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026067.3 | NP_080343.4 | 3 | 592 | ||
GenBank mRNA | AK014041.1 | - | 3 | 593 | ||
AK014410.1 | - | 3 | 594 | |||
AK144223.1 | - | 3 | 581 | |||
AK165026.1 | - | 3 | 521 | |||
AK172005.1 | - | 3 | 559 | |||
BC046412.1 | - | 3 | 540 | 5354985 |