Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2410127L17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026120.4 | NP_080396.2 | 6, 7 | 1216 | ||
GenBank mRNA | AK010784.1 | - | 6, 7 | 1101 | ||
BC019559.1 | - | 6, 7 | 1406 | |||
BC043335.1 | - | 6, 7 | 1170 | 5030742 | ||
BC092281.1 | - | 6, 7 | 1048 | 4950426 |