Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fundc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026126.4 | NP_080402.3 | 4, 5 | 409 | ||
GenBank mRNA | AK013460.1 | - | 4, 5 | 387 | ||
AK014705.1 | - | 4, 5 | 386 | |||
AK031214.1 | - | 4, 5 | 386 | |||
AY234858.1 | - | 4, 5 | 375 | |||
BC042794.1 | - | 3, 4 | 366 | |||
BC049566.1 | - | 4, 5 | 384 | 6742356 | ||
BC061464.1 | - | 4, 5 | 379 | 6819539 |