Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023343.2 | NP_075832.1 | 9 | 872 | ||
GenBank mRNA | AF117831.1 | - | 4 | 326 | ||
AK003007.1 | - | 8 | 1845 | |||
AK005408.1 | - | 9 | 895 | |||
AK019158.1 | - | 3 | 256 | |||
BC011427.1 | - | 9 | 842 | |||
BC026953.1 | - | 9 | 872 | 5133980 | ||
BC027439.1 | - | 9 | 761 | 4164301 |