Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026164.2 | NP_080440.2 | 6, 7 | 1849 | ||
GenBank mRNA | AB044139.1 | - | 6, 7 | 1728 | ||
AK004621.1 | - | 6, 7 | 1816 | |||
AK084928.1 | - | 1516 | ||||
AK145776.1 | - | 6, 7 | 1759 | |||
AK163211.1 | - | 6, 7 | 1817 | |||
AK168475.1 | - | 6, 7 | 1812 | |||
BC019364.1 | - | 3, 4 | 303 | |||
BC127056.1 | - | 5, 6 | 1709 | 40109753 |