Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026231.2 | NP_080507.2 | 2 | 597 | ||
GenBank mRNA | AK020021.1 | - | 1 | 207 | ||
AK029724.1 | - | 2 | 596 | |||
AK033409.1 | - | 2 | 635 | |||
AK037169.1 | - | 2 | 636 | |||
AK044441.1 | - | 2 | 605 | |||
BC047136.1 | - | 2 | 597 | 6311866 | ||
BC061240.1 | - | 2 | 593 | 6743615 |