Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyx1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163725.1 | NP_001157197.1 | 6 | 852 | ||
NM_026314.3 | NP_080590.3 | 8 | 1212 | |||
GenBank mRNA | AK005832.1 | - | 5 | 662 | ||
AK015345.1 | - | 4 | 391 | |||
AY428507.1 | - | 8 | 1212 | |||
AY464142.1 | - | 6 | 852 | |||
BC026462.1 | - | 8 | 1169 | 4483015 |