Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026319.3 | NP_080595.2 | 20 | 1893 | ||
GenBank mRNA | AK006492.1 | - | 11 | 1000 | ||
CD740935.1 | - | 1 | 261 |