Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026384.3 | NP_080660.1 | 5 | 780 | ||
GenBank mRNA | AF384160.1 | - | 5 | 638 | ||
AK002443.1 | - | 5 | 785 | |||
AK016660.1 | - | 5 | 729 | |||
BC043447.1 | - | 5 | 780 | 5125951 |