Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprgl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026388.2 | NP_080664.1 | 3 | 614 | ||
GenBank mRNA | AK004324.1 | - | 2 | 227 | ||
AK004777.1 | - | 3 | 597 | |||
AK155413.1 | - | 3 | 609 | |||
AK195680.1 | - | 1 | 64 | |||
AK196787.1 | - | 1 | 64 | |||
AK206032.1 | - | 1 | 64 | |||
AK207196.1 | - | 1 | 64 | |||
AK209255.1 | - | 1 | 64 | |||
AK210976.1 | - | 1 | 64 | |||
AK211127.1 | - | 1 | 64 | |||
AK213307.1 | - | 1 | 64 | |||
AK213716.1 | - | 2 | 163 | |||
AK213906.1 | - | 2 | 162 | |||
AK218094.1 | - | 1 | 64 | |||
AK219515.1 | - | 2 | 163 | |||
AK220054.1 | - | 1 | 64 | |||
BC019544.1 | - | 3 | 578 | 4211201 | ||
BY739996.1 | - | 3 | 613 |