Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Saraf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026432.3 | NP_080708.3 | 3 | 701 | ||
GenBank mRNA | AK007787.1 | - | 3 | 628 | ||
AK075744.1 | - | 3 | 592 | |||
AK077118.1 | - | 3 | 688 | |||
AK088728.1 | - | 3 | 584 | |||
AK093942.1 | - | 3 | 676 | |||
AK163218.1 | - | 3 | 634 | |||
AK178694.1 | - | 1 | 141 | |||
AK211246.1 | - | 2 | 283 | |||
AK213348.1 | - | 1 | 141 | |||
AK213493.1 | - | 2 | 318 | |||
BC013497.1 | - | 3 | 642 | 4222385 | ||
BC022616.1 | - | 3 | 679 | 4187981 | ||
BC024888.1 | - | 3 | 637 | 3988822 |