Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026487.3 | NP_080763.2 | 2 | 89 | ||
GenBank mRNA | AK014967.1 | - | 2 | 106 | ||
AK030719.1 | - | 2 | 109 | |||
AK033639.1 | - | 2 | 113 | |||
AK044925.1 | - | 2 | 89 | |||
AK075685.1 | - | 2 | 133 | |||
AK150469.1 | - | 2 | 109 | |||
AK152059.1 | - | 2 | 109 | |||
AK165953.1 | - | 2 | 124 | |||
BC029085.1 | - | 2 | 89 | 3593451 | ||
BC043051.1 | - | 2 | 74 | 5708377 |