Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080556.3 | NP_542123.3 | 6 | 871 | ||
GenBank mRNA | AK010325.1 | - | 6 | 828 | ||
AK030340.1 | - | 6 | 801 | |||
AK048012.1 | - | 6 | 790 | |||
AK050338.1 | - | 6 | 818 | |||
AK075720.1 | - | 6 | 871 | |||
AK088905.1 | - | 6 | 843 | |||
AK159757.1 | - | 6 | 816 | |||
AK209166.1 | - | 2 | 204 | |||
AK212229.1 | - | 2 | 206 | |||
BC003862.1 | - | 6 | 810 | 3488326 |