Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026554.4 | NP_080830.1 | 2 | 315 | ||
GenBank mRNA | AK008980.1 | - | 2 | 255 | ||
AK012659.1 | - | 2 | 235 | |||
AK014016.1 | - | 2 | 234 | |||
AK077841.1 | - | 2 | 223 | |||
AK078181.1 | - | 2 | 246 | |||
AK135597.1 | - | 2 | 229 | |||
AK147027.1 | - | 2 | 247 | |||
AK177380.1 | - | 1 | 90 | |||
BC118926.2 | - | 2 | 223 | 40090166 | ||
BY035864.1 | - | 2 | 349 |