Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026641.2 | NP_080917.1 | 17 | 2084 | ||
GenBank mRNA | AK019542.1 | - | 4 | 2001 | ||
AK042231.1 | - | 17 | 2009 | |||
AK147305.1 | - | 16 | 5611 | |||
BC013814.1 | - | 12 | 1373 | |||
BC034101.1 | - | 17 | 1979 | 4501451 |