Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005846.2 | NP_001005846.1 | 11 | 1372 | ||
NM_026656.4 | NP_080932.2 | 11 | 1400 | |||
GenBank mRNA | AF503575.1 | - | 11 | 1389 | ||
AK014467.1 | - | 11 | 1303 | |||
AK019454.1 | - | 11 | 1269 | |||
AK150446.1 | - | 11 | 1368 | |||
AK150547.1 | - | 11 | 1368 | |||
AK152067.1 | - | 11 | 1368 | |||
AK152943.1 | - | 11 | 1368 | |||
AK153290.1 | - | 11 | 1368 | |||
AK159172.1 | - | 11 | 1400 | |||
AY083532.1 | - | 10 | 1218 | |||
BC029847.1 | - | 11 | 1249 | 3967456 |