Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024222.2 | NP_077184.2 | 11 | 1976 | ||
GenBank mRNA | AK012153.1 | - | 11 | 1646 | ||
AK018758.2 | - | 11 | 1686 | |||
AK145674.1 | - | 11 | 1617 | |||
AK148065.1 | - | 11 | 1675 | |||
AK152899.1 | - | 11 | 2024 | |||
AK154979.1 | - | 11 | 1665 | |||
AK170079.1 | - | 11 | 1688 | |||
AX799084.1 | - | 11 | 1687 | |||
BC003206.1 | - | 6 | 688 | |||
BC013054.1 | - | 1 | 94 | |||
BC052433.1 | - | 11 | 1976 | 6837097 |