Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrriq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290510.1 | NP_001277439.1 | 6 | 1970 | ||
NM_026668.2 | NP_080944.2 | 8 | 2133 | |||
GenBank mRNA | AK015024.1 | - | 6 | 1921 | ||
AK015586.1 | - | 2 | 145 | |||
AK019775.1 | - | 8 | 2134 | |||
BC145806.1 | - | 8 | 1834 | 40131109 |