Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291235.1 | NP_001278164.1 | 2 | 111 | ||
NM_001291236.1 | NP_001278165.1 | 2 | 587 | |||
NM_001291237.1 | NP_001278166.1 | 3 | 295 | |||
NM_026708.2 | NP_080984.1 | 2 | 422 | |||
GenBank mRNA | AA671983.1 | - | 3 | 312 | ||
AK002265.1 | - | 2 | 370 | |||
AK002703.1 | - | 2 | 318 | |||
AK046577.1 | - | 3 | 2302 | |||
AK161721.1 | - | 322 | ||||
AK165432.1 | - | 534 | ||||
BC005702.1 | - | 2 | 314 | 3598444 | ||
BC039182.1 | - | 2 | 557 | 3500908 | ||
BG083251.2 | - | 2 | 169 |