Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026775.4 | NP_081051.1 | 3 | 418 | ||
GenBank mRNA | AK003676.1 | - | 3 | 401 | ||
AK084117.1 | - | 3 | 401 | |||
AK150605.1 | - | 3 | 390 | |||
AK152643.1 | - | 3 | 401 | |||
AK152656.1 | - | 3 | 390 | |||
AK186798.1 | - | 3 | 163 | |||
AK191153.1 | - | 1 | 21 | |||
AK198152.1 | - | 1 | 21 | |||
AK201939.1 | - | 1 | 21 | |||
AK206502.1 | - | 1 | 21 | |||
AK207697.1 | - | 1 | 21 | |||
AK216393.1 | - | 1 | 21 | |||
BC006774.1 | - | 1 | 73 | |||
BC064755.1 | - | 3 | 392 | 30029180 | ||
BC085097.1 | - | 3 | 397 | 5349826 |