Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080575.2 | NP_542142.1 | 5 | 877 | ||
GenBank mRNA | AB046742.1 | - | 5 | 866 | ||
AK038517.1 | - | 1 | 589 | |||
AK088244.1 | - | 5 | 856 | |||
AK146568.1 | - | 5 | 878 | |||
AK158119.1 | - | 5 | 872 | |||
AK172412.1 | - | 5 | 863 | |||
AK172470.1 | - | 5 | 852 | |||
AK173275.1 | - | 5 | 838 | |||
BC030930.1 | - | 1 | 51 | 4911679 | ||
BC092278.1 | - | 5 | 906 | 3710505 |