Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145453.2 | NP_663428.2 | 3 | 617 | ||
GenBank mRNA | AK044802.1 | - | 3 | 479 | ||
AK051610.1 | - | 3 | 626 | |||
BC010687.1 | - | 2 | 532 | |||
BC024496.1 | - | 2 | 541 | 4982244 |