Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080557.2 | NP_542124.1 | 14 | 1322 | ||
GenBank mRNA | AK007706.1 | - | 14 | 1363 | ||
AK030657.1 | - | 14 | 1335 | |||
AK031368.1 | - | 1334 | ||||
AK135356.1 | - | 14 | 1329 | |||
BC006043.1 | - | 14 | 1312 | |||
BC016599.1 | - | 14 | 1322 | 2644991 |