Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178377.4 | NP_848464.1 | 3, 4 | 260 | ||
GenBank mRNA | AB052770.1 | - | 3, 4 | 273 | ||
AK139321.1 | - | 276 | ||||
AK159099.1 | - | 3, 4 | 276 | |||
AK166907.1 | - | 3, 4 | 274 | |||
BC034879.1 | - | 3, 4 | 260 | 3468632 |