Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175150.3 | NP_780359.2 | 3, 4 | 829 | ||
GenBank mRNA | AK079517.1 | - | 3, 4 | 776 | ||
AK160051.1 | - | 3, 4 | 798 | |||
AK167701.1 | - | 3, 4 | 737 | |||
AK167838.1 | - | 3, 4 | 795 | |||
AK167977.1 | - | 3, 4 | 794 | |||
BC029015.1 | - | 2, 3 | 565 | |||
BC038082.1 | - | 1, 2 | 87 | |||
BC062180.1 | - | 3, 4 | 829 | 6336176 | ||
BC094275.1 | - | 3, 4 | 665 |