Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mif4gd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243584.1 | NP_001230513.1 | 6 | 749 | ||
NM_001243586.1 | NP_001230515.1 | 5 | 976 | |||
NM_001243587.1 | NP_001230516.1 | 6 | 692 | |||
NM_027162.4 | NP_081438.4 | 5 | 1033 | |||
GenBank mRNA | AK003708.1 | - | 810 | |||
AK010175.1 | - | 6 | 703 | |||
AK150749.1 | - | 6 | 703 | |||
BC019515.1 | - | 6 | 660 | 4460171 | ||
BC026740.1 | - | 6 | 709 | 4015105 | ||
BC026745.1 | - | 6 | 519 | |||
BC055812.1 | - | 5 | 1033 | 4934728 | ||
BQ174854.1 | - | 1 | 636 |