Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027187.3 | NP_081463.1 | 3 | 291 | ||
GenBank mRNA | AK010292.1 | - | 3 | 281 | ||
AK146518.1 | - | 3 | 292 | |||
BC029252.1 | - | 3 | 321 | 3666709 | ||
BC038158.1 | - | 3 | 365 | 1493985 |